Order Entry
Austria
ContactUsLinkComponent
Test paper, qualitative, Nitratesmo
Test paper, qualitative, Nitratesmo
Catalog # MANA90611
Supplier:  MACHEREY-NAGEL
CAS Number:  
undefined
Test paper, qualitative, Nitratesmo
Catalog # MANA90611
Supplier:  MACHEREY-NAGEL
CAS Number:  

Specifications

  • Chemical method:
    Imidazoline
  • Pk:
    1 Roll
  • Description:
    Nitratesmo test papers (nitrate and nitrite)

Specifications

About this item

This test paper allows the reliable detection of nitrate and nitrite. It shows different colours for the two ions.

  • Colour change: Nitrate: White to red
  • Nitrite: White to yellow

Limit of sensitivity: 10 mg/L Nitrate (NO³⁻), 5 mg/L Nitrite (NO²⁻).

Packaging: Reel of 5 m length and 10 mm width.