Order Entry
Austria
ContactUsLinkComponent
Test paper, qualitative, dipyridyl paper for iron II ions
Test paper, qualitative, dipyridyl paper for iron II ions
Catalog # MANA90725
Supplier:  MACHEREY-NAGEL
CAS Number:  
undefined
Test paper, qualitative, dipyridyl paper for iron II ions
Catalog # MANA90725
Supplier:  MACHEREY-NAGEL
CAS Number:  

Specifications

  • Pk:
    200
  • Description:
    Dipyridyl paper, for (Fe²⁺) ions

Specifications

About this item

The test paper allows the sensitive and specific detection of Fe(II) ions. Even small amounts of Fe(II) can safely be detected in presence of large amounts of Fe(III).

  • Colour changes: White to red

Limit of sensitivity: 2 mg/L Fe²⁺