Order Entry
Austria
ContactUsLinkComponent
Test paper, qualitative, Cuprotesmo
Test paper, qualitative, Cuprotesmo
Catalog # MANA90601
Supplier:  MACHEREY-NAGEL
CAS Number:  
undefined
Test paper, qualitative, Cuprotesmo
Catalog # MANA90601
Supplier:  MACHEREY-NAGEL
CAS Number:  

Specifications

  • Chemical method:
    Biquinoline
  • Pk:
    40
  • Description:
    Cuprotesmo test paper

Specifications

About this item

This test paper allows the quick and easy detection of Copper(I) and Copper(II) ions.

  • Colour change: Yellow-white to pink-purple

It used for the detection of copper and copper salts on surfaces and in ash, for the detection of pores in metallic coatings on copper containing materials, in criminal trace investigations (bullets), and for the detection of copper-containing pesticides on fruit and vegetables. The test paper is specific for copper.

Limit of sensitivity:
0,05 μg on surfaces
3 to 5 mg/L Cu in solutions

Packaging: Pack of 40 sheets 40×25 mm