
Tetrabutylammonium hydrogen sulphate, HiPerSolv CHROMANORM® for HPLC
85794.150
:
- Pack type:Plastic bottle
- Pk:25 g
- Seq:3308
- Assay (Acidimetric):≥ 99,00 %
- IR Spectrum:Passes test
- Absorbance (200 nm) (0.005 mol/l):≤ 0,155
- Absorbance (220 nm) (0.005 mol/l):≤ 0,046
- Absorbance (250 nm) (0.005 mol/l):≤ 0,009
:
:339.54 g/mol
:169…172 °C
:1.01 g/cm³ (25 °C)
:251-068-5
:GHS07
:32503-27-8
:(C₄H₉)₄NHSO₄
:MFCD00011637