Order Entry
Austria
ContactUsLinkComponent
 
Neocarmine W "Fesago" colour reagent, Sigma-Aldrich®
Catalog # 1.06732.1000
Supplier:  Merck
CAS Number:  
Neocarmine W "Fesago" colour reagent, Sigma-Aldrich®
Catalog # 1.06732.1000
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Suitability for coloration of textiles:
    passes test

Specification Test Results

About this item

 :
Density:1.01 g/cm3 (20 °C)
Storage Temperature:Ambient