Order Entry
Austria
ContactUsLinkComponent
Pesticide reference standard, Atrazines, Honeywell Fluka™
Pesticide reference standard, Atrazines, Honeywell Fluka™
Certificates
Pesticide reference standard, Atrazines, Honeywell Fluka™

Specifications

  • CAS No.:
    1912-24-9
  • Formula:
    C₈H₁₄ClN₅
  • Storage temperature:
    Ambient
  • Description:
    Atrazine

Specifications

Frequently Bought Together