Order Entry
Austria
ContactUsLinkComponent
Pharmaceutical analytical standard, Nirvanol-d5, reference material, Honeywell Fluka™
Pharmaceutical analytical standard, Nirvanol-d5, reference material, Honeywell Fluka™
Catalog # HONCR1515-10MG
CAS Number:  
Pharmaceutical analytical standard, Nirvanol-d5, reference material, Honeywell Fluka™
Catalog # HONCR1515-10MG
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mg
  • CAS No.:
    119458-27-4
  • Description:
    Nirvanol-d₅
  • Formula:
    C₁₁H₇D₅N₂O₂
  • Storage temperature:
    2...8 °C
  • Description:
    Nirvanol-d₅
  • Freezing point:
    186...189 °C

Specifications

About this item

High quality inorganic and organic reference materials produced according to ISO 9001.

  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results