Order Entry
Austria
ContactUsLinkComponent
ASTM® D6584 individual stock and internal standards mix Kit, analytical standard, Honeywell Fluka™
ASTM® D6584 individual stock and internal standards mix Kit, analytical standard, Honeywell Fluka™
Catalog # HONCR1386-1EA
CAS Number:  
ASTM® D6584 individual stock and internal standards mix Kit, analytical standard, Honeywell Fluka™
Catalog # HONCR1386-1EA
CAS Number:  

Specifications

  • Pk:
    1
  • Description:
    ASTM® D6584 Individual Stock and Internal Standards Mix Kit
  • Storage temperature:
    2...8 °C
  • Description:
    ASTM® D6584 Individual Stock and Internal Standards Mix Kit
  • Pk Info:
    1 kit

Specifications

About this item

High quality inorganic and organic reference materials produced according to ISO 9001.

  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results

Kit contains:
R1380-1ML ASTM® D6584 Glycerin solution, 500 μg/ml in pyridine, reference material
R1381-3ML ASTM® D6584 Monoolein solution, 5000 μg/ml in pyridine, reference material
R1382-2ML ASTM® D6584 1,3-Diolein solution, 5000 μg/ml in pyridine, reference material
R1383-2ML ASTM® D6584 Triolein solution, 5000 μg/ml in pyridine, reference material
R1384-5ML ASTM® D6584 Butanetriol solution, 1000 μg/ml in pyridine, reference material
R1385-5ML ASTM® D6584 Tricaprin solution, 8000 μg/ml in pyridine, reference material