Order Entry
Austria
ContactUsLinkComponent
 
ASTM® D6584 Monoolein solution, 5000 μg/ml in pyridine, analytical standard, Honeywell Fluka™
  HONCR1381-3ML
 :  
ASTM® D6584 Monoolein solution, 5000 μg/ml in pyridine, analytical standard, Honeywell Fluka™
  HONCR1381-3ML
 :  

 

  • Concentration:
    5000 μg/ml in pyridine
  • Pack type:
    Glass bottle
  • Pk:
    3 mL
  • Description:
    ASTM® D6584 Monoolein solution
  • Storage temperature:
    2...8 °C
  • Description:
    ASTM® D6584 Monoolein solution
  • Flash point:
    20 °C
  • Boiling point:
    115 °C
  • Freezing point:
    –42 °C

 

 

High quality inorganic and organic reference materials produced according to ISO 9001.

  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results