Pesticide reference standard, Dimethirimol, neat, Honeywell Fluka™
Catalog # HONCR1307-50MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:50 mg
- CAS No.:5221-53-4
- Description:Dimethirimol
- Environmentally Preferable:
- Formula:C₁₁H₁₉N₃O
- Storage temperature:2...8 °C
- Description:Dimethirimol
- Freezing point:102 °C
Specifications
About this item
High quality inorganic and organic reference materials produced according to ISO 9001.
- Ensure you have the information you need to quickly start your analysis, and pass relevant audits
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results