Order Entry
Austria
ContactUsLinkComponent
Gram's decolourising solution contains acetone and ethanol
Gram's decolourising solution contains acetone and ethanol
Catalog # 9761.5000
Supplier:  VWR Chemicals
CAS Number:  
undefined
Gram's decolourising solution contains acetone and ethanol
Catalog # 9761.5000
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pk:
    5 L
  • Seq:
    5190

Specifications

Specification Test Results

  • Appearance:
    Clear liquid

Specification Test Results

About this item

 :
 :37348-17-7
MDL Number:MFCD00240361