

Gram's decolourising solution for Gram staining, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
:37348-17-7
MDL Number:MFCD00240361
Density:0.79 g/cm3 (20 °C)
UN:1993
ADR:3,
:M-Clarity™ quality level = MQ300
Flash Pt:–10 °C
:GHS02
:GHS07
:1
Specification Test Results
- Suitability for microscopy:passes test
- Staphylococcus aureus:dark blue-violet
- E.coli:pink to red
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERC1.10218.0500_TDS_EN
Product specification
Merck The Color of Tissue Diagnostics
Product Literature
msds_110218
SDS
Frequently Bought Together