Order Entry
Austria
ContactUsLinkComponent
Gram's decolourising solution contains acetone and 2-propanol
Gram's decolourising solution contains acetone and 2-propanol
Catalog # 99502.5000
Supplier:  VWR Chemicals
CAS Number:  
undefined
Gram's decolourising solution contains acetone and 2-propanol
Catalog # 99502.5000
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pk:
    5 L
  • Seq:
    5190

Specifications

Specification Test Results

  • Solution 50/50 (V/V):
    Clear liquid

Specification Test Results

About this item

 :
UN:1993
ADR:3,II
 :GHS02
 :37348-17-7
MDL Number:MFCD00240361