Orders LinkContactUsLinkComponent
Hydrometers
Supplier:   GERING
Hydrometers
Supplier:  GERING
Certificates
Hydrometers
9 Options Available Below
Product Details & Documents
Product Family Options
Product Information
  • Range
    Division
    Availability
    Price
  • 0,900 - 1,000 g/ml
    0,001 g/ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 58,30 €
    58,30 €
    1 * 1 ST
       58,30 €
  • 1,000 - 1,100 g/ml
    0,001 g/ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 58,60 €
    58,60 €
    1 * 1 ST
       58,60 €
  • 1,000 - 1,200 g/ml
    0,002 g/ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 61,40 €
    61,40 €
    1 * 1 ST
       61,40 €
  • 1,000 - 1,500 g/ml
    0,005 g/ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 70,30 €
    70,30 €
    1 * 1 ST
       70,30 €
  • 1,100 - 1,200 g/ml
    0,001 g/ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 58,60 €
    58,60 €
    1 * 1 ST
       58,60 €
  • 1,200 - 1,300 g/ml
    0,001 g/ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 58,60 €
    58,60 €
    1 * 1 ST
       58,60 €
  • 1,200 - 1,400 g/ml
    0,002 g/ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 61,40 €
    61,40 €
    1 * 1 ST
       61,40 €
  • 1,300 - 1,400 g/ml
    0,001 g/ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 63,70 €
    63,70 €
    1 * 1 ST
       63,70 €
  • 1,400 - 1,500 g/ml
    0,001 g/ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 63,70 €
    63,70 €
    1 * 1 ST
       63,70 €