ContactUsLinkComponent
Deuterium lamp for UV/Vis detector
Deuterium lamp for UV/Vis detector
Catalog # PERCN2920149
Supplier:  PerkinElmer
Deuterium lamp for UV/Vis detector
Catalog # PERCN2920149
Supplier:  PerkinElmer
Specifications
  • For:
    LC 200/785A
  • Description:
    Deuterium lamp
  • Pk:
    1
Specifications
About this item
Deuterium lamp for the LC series 200 UV/Vis detector, provide outstanding performance and lamp life.
Lamp changes are quick and easy due to a unique, self-aligning lamp mount.