Recirculating chiller, SONICATOR®
High capacity recirculating chiller, 115 - 230 V, 50/60 Hz, 5...45 °C, 0,4 kW, 320×280×320 mm
Catalog # QSON4905-220
Supplier: QSONICA LLC
Jump to:
Product Details & Documents
Sonication generates heat which may be detrimental to some applications. Attempting to control temperature with ice and/or repeatedly changing out water is tedious and no longer necessary. This chiller automates the sample cooling process.
- Quick-connect tubing and fittings attach the chiller to the ports on the cup horn or microplate horn
- When used in conjunction with the pulsed sonication mode, desired water temperature will be maintained
Older model cup horns may require special fittings.
Ordering information: Quick-connect tubing and fittings should be ordered separately.
Specifications
- Temperature Range:5...45 °C
- Description:High capacity recirculating chiller, 115 - 230 V, 50/60 Hz
- W×D×H:320×280×320 mm
- Cooling capacity:0,4 kW
Accessories
Product Information
- Catalog #ForAvailabilityPrice
- Cup horn tubing and connector set, includes 2 male fittings, 2 right angle male fittings and 2 female fittingsCup horn tubing and connector set, includes 2 male fittings, 2 right angle male fittings and 2 female fittingsQSON4911High capacity recirculating chillerLog in for availability1 * 1 ST - 256,00 €
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



