Lids for 96-well PCR plates, ComfortLid
HAMR814300
- Sterility:Sterile
- Colour:Clear
- Description:Lid for 96-well PCR plate, ComfortLid
- Environmentally Preferable:
- Pk:50
Gentle pressure is applied to the ComfortLid to seal PCR plates and prevent evaporation and contamination, used in equipped liquid handling workstations.
- Free of DNase, RNase, pyrogen (endotoxins), human DNA, PCR inhibitors
- Made of polycarbonate (PC) and thermoplastic elastomers (TPE)
These cost-effective and disposable lid has a stackable design that can be transported by all robotic transport arms.
: Packed as 10 lids/blister and 5 blister/box.