Order Entry
Austria
ContactUsLinkComponent
Epidermal growth factor, recombinant, human, Calbiochem®
Epidermal growth factor, recombinant, human, Calbiochem®
Catalog # 324831-200
CAS Number:  
Epidermal growth factor, recombinant, human, Calbiochem®
Catalog # 324831-200
CAS Number:  

Specifications

  • Pk:
    200 µG
  • Description:
    Epidermal growth factor (EGF), recombinant, human
  • Formulation:
    Lyophilised from sterile-filtered PBS

Specifications

About this item

Recombinant, human epidermal growth factor is a protein expressed in E. coli which is identical to human EGF except for an additional N-terminal methionine.

  • Purity: ≥95% by SDS-PAGE
  • Store at –20 °C

It is mitogenic for a wide range of ectoderm- and endoderm-derived cells from tissue such as breast, cornea, epidermis, dermis, liver, pancreas, nerve, amnion, and adrenal medulla. Acts as a survival factor in preventing apoptosis.

Packaging: Packed in plastic ampoule.