Order Entry
Austria
ContactUsLinkComponent
Test papers, qualitative, cobalt
Test papers, qualitative, cobalt
Catalog # MANA90728
Supplier:  MACHEREY-NAGEL
CAS Number:  
undefined
Test papers, qualitative, cobalt
Catalog # MANA90728
Supplier:  MACHEREY-NAGEL
CAS Number:  

Specifications

  • Pk:
    100
  • Description:
    Cobalt ions (Co²⁺)
  • Range:
    25 mg/l (Co²⁺)

Specifications

About this item

This test paper allows the quick and easy detection of Cobalt (Co²⁺). For non-destructive testing of materials apply a drop of acid solution (50 ml hydrogen peroxide 3% + 7,5 ml O-phosphoric acid 85% + 5 ml hydrochloric acid 37 %) to the degreased surface.

  • Colour change: White to blue

After 30 to 60 seconds absorb the liquid with the test paper. In the presence of Cobalt a blue colour appears. The intensity of the colour varies according to the concentration. With this procedure Cobalt contents >0,5% are safely detected.

Limit of sensitivity: 25 mg/l Co2+

Delivery: Pack of 100 strips 20×70 mm