Order Entry
Austria
Orders LinkContactUsLinkComponent
Test papers, qualitative, Bismuth

Test papers, qualitative, Bismuth

Bismuth Test paper for Bismuth, 60 - mg/L - Bi³⁺, Complexation with a quinoline, 48×25×84 mm (package), 200 test(s), 200
Catalog # MANA90733
Supplier:   MACHEREY-NAGEL
Test papers, qualitative, Bismuth
Catalog # MANA90733
Supplier:  MACHEREY-NAGEL
CAS Number:  
Test papers, qualitative, Bismuth
Test papers, qualitative, Bismuth

Bismuth Test paper for Bismuth, 60 - mg/L - Bi³⁺, Complexation with a quinoline, 48×25×84 mm (package), 200 test(s), 200

CAS Number:  
Price
46,40 €
Log in for availability
1 * 200 ST
Product Details & Documents

This test paper allows the rapid detection of Bismuth. In the presence of Bismuth ions (Bi³⁺) it shows an orange-red discolouration on a pale-yellow background.

  • Colour change: Pale-yellow to orange-red

Limit of sensitivity: 60 mg/L Bi³⁺ (Nitric acid >3% leads to reduced sensitivity)

 : Pack of 200 strips 20×70 mm.

Specifications

Specifications

  • Chemical method:
    Complexation with a quinoline
  • Pk:
    200
  • Description:
    Bismuth Test paper for Bismuth
  • Dimensions:
    48×25×84 mm (package)
  • Tests per kit:
    200 test(s)
  • Range:
    60 - mg/L - Bi³⁺
  • Cat. No.:
    MANA90733
  • Sensitivity:
    Bismuth - 60 mg/L Bi³⁺
  • Contents:
    200 test strips in a plastic box
  • Platform:
    Qualitative test paper
  • Compliance:
    Not CE certified and no hazardous material
  • Shelf life (months):
    24 (from production)
  • W×D×H:
    20×70 mm (net)
  • Method:
    Bismuth - pale-yellow → orange-red
  • Weight:
    57 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.