VISOCOLOR® Colorimetric Test, Cyanuric acid
Supplier: MACHEREY-NAGEL
Certificates
About this item
Reagents for the determination of cyanuric acid in water samples.
- Combines simple implementation with high measurement reliability
Simple determination directly on site or in the laboratory.
Delivery: Remark: Measuring range on PF-12Plus. Range on other photometers can be different.
Specifications
- Model:MANA931223
- Platform:Turbidity-based test kits
- Storage temperature:15…25 °C
- Shelf life (months):18
- Weight:86 g (including packaging)
Specifications
Related Information
Frequently Bought Together