VISOCOLOR® Colorimetric Test, Iron
Supplier: MACHEREY-NAGEL
About this item
Reagents for the determination of iron in water samples.
- Combines simple implementation with high measurement reliability
Simple determination directly on site or in the laboratory.
6 Options Available Below
Product Details & Documents
Reagents for the determination of iron in water samples.
- Combines simple implementation with high measurement reliability
Simple determination directly on site or in the laboratory.
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- MANA931026
- MANA920040
- MANA931025
- MANA931226
- MANA920140
- MANA931225
- Model:
- MANA931026
- MANA920040
- MANA931025
- MANA931226
- MANA920140
- MANA931225
- Platform:
- Colorimetric test kits
- Colorimetric test kits
- Colorimetric test kits
- Colorimetric test kits
- Colorimetric test kits
- Colorimetric test kits
- Storage temperature:
- 15...25 °C
- 15...25 °C
- 15...25 °C
- 15...25 °C
- 15...25 °C
- 15...25 °C
- Shelf life (months):
- 24
- 24
- 24
- 24
- 24
- 24
- Weight:
- 189,6 g (Including packaging)
- 414 g (Including packaging)
- 190,59 g (Including packaging)
- 101 g (Including packaging)
- 280,4 g (Including packaging)
- 98,6 g (Including packaging)
Specifications
Product Family Options
Test kits
Product Information
- ContentsRangeChemical methodFor unitDimensionsTypeTests per kitPkAvailabilityPrice
- MANA931026VISOCOLOR® ECO Iron 2, test no. 5-26. Measuring range on PF-12Plus. Range on other photometers can be differentVISOCOLOR® ECO Iron 2, test no. 5-26. Measuring range on PF-12Plus. Range on other photometers can be differentReagents for 100 tests, color chart and accessories in a box0 • 0,04 • 0,07 • 0,10 • 0,15 • 0,20 • 0,30 • 0,50 • 1,0 mg/L (Fe); 0,04 - 2,00 mg/L (Fe)TriazinePF-12Plus;PF-3 Drinking Water;Advance;PF-3 Pool;Visual168×112×40 mm (Including packaging)Sea water compatible; Hazardous material: No1001 KITLog in for availability1 * 1 KIT59,50 €Specifications:
More
Specifications
Cat. No.MANA931026ModelMANA931026PlatformColorimetric test kitsStorage temperature15...25 °CShelf life (months)24Weight189,6 g (Including packaging) - MANA920040VISOCOLOR® HE IronVISOCOLOR® HE IronReagents for 300 tests, color chart and accessories in a box0,0 • 0,01 • 0,02 • 0,03 • 0,04 • 0,05 • 0,07 • 0,10 • 0,15 • 0,20 mg/L (Fe)TriazineVisual240×135×46 mm (Including packaging)Sea water non compatible; Hazardous material: No3001 KITLog in for availability1 * 1 KIT124,00 €Specifications:
More
Specifications
Cat. No.MANA920040ModelMANA920040PlatformColorimetric test kitsStorage temperature15...25 °CShelf life (months)24Weight414 g (Including packaging) - MANA931025VISOCOLOR® ECO Iron 1, test no. 5-25. Measuring range on PF-12Plus. Range on other photometers can be differentVISOCOLOR® ECO Iron 1, test no. 5-25. Measuring range on PF-12Plus. Range on other photometers can be differentReagents for 200 tests, color chart and accessories in a box0 • 0,04 • 0,07 • 0,10 • 0,15 • 0,20 • 0,30 • 0,50 • 1,0 mg/L (Fe); 0,04 - 2,00 mg/L (Fe)TriazinePF-12Plus;PF-3 Fish;PF-3 Pool;PF-3 Drinking Water;Advance;Visual168×112×40 mm (Including packaging)Sea water compatible; Hazardous material: Yes200200 TestsLog in for availability1 * 200 Tests74,70 €Specifications:
More
Specifications
Cat. No.MANA931025ModelMANA931025PlatformColorimetric test kitsStorage temperature15...25 °CShelf life (months)24Weight190,59 g (Including packaging)
Refill packs
Product Information
- ContentsRangeChemical methodFor unitDimensionsTypeTests per kitPkAvailabilityPrice
- MANA931226VISOCOLOR® ECO Iron 2, refill pack, test no. 5-26. Measuring range on PF-12Plus. Range on other photometers can be differentVISOCOLOR® ECO Iron 2, refill pack, test no. 5-26. Measuring range on PF-12Plus. Range on other photometers can be differentReagents for 100 tests in a box0 • 0,04 • 0,07 • 0,10 • 0,15 • 0,20 • 0,30 • 0,50 • 1,0 mg/L (Fe); 0,04 - 2,00 mg/L (Fe)TriazinePF-12Plus;PF-3 Fish;PF-3 Pool;PF-3 Drinking Water;Advance;Visual170×115×70 mm (Including packaging)Sea water compatible; Hazardous material: No1001 KITLog in for availability1 * 1 KIT45,00 €Specifications:
More
Specifications
Cat. No.MANA931226ModelMANA931226PlatformColorimetric test kitsStorage temperature15...25 °CShelf life (months)24Weight101 g (Including packaging) - MANA920140VISOCOLOR® HE Iron , refill packVISOCOLOR® HE Iron , refill packReagents for 300 tests in a box0,0 • 0,01 • 0,02 • 0,03 • 0,04 • 0,05 • 0,07 • 0,10 • 0,15 • 0,20 mg/L (Fe)TriazineVisual170×115×70 mm (Including packaging)Sea water non compatible; Hazardous material: No3001 KITLog in for availability1 * 1 KIT76,80 €Specifications:
More
Specifications
Cat. No.MANA920140ModelMANA920140PlatformColorimetric test kitsStorage temperature15...25 °CShelf life (months)24Weight280,4 g (Including packaging) - MANA931225VISOCOLOR® ECO Iron 1, refill pack, test no. 5-25. Measuring range on PF-12Plus. Range on other photometers can be differentVISOCOLOR® ECO Iron 1, refill pack, test no. 5-25. Measuring range on PF-12Plus. Range on other photometers can be differentReagents for 200 tests in a box0 • 0,04 • 0,07 • 0,10 • 0,15 • 0,20 • 0,30 • 0,50 • 1,0 mg/L (Fe); 0,04 - 2,00 mg/L (Fe)TriazinePF-12Plus;PF-3 Fish;PF-3 Pool;PF-3 Drinking Water;Advance;Visual170×115×70 mm (Including packaging)Sea water compatible; Hazardous material: Yes2001Log in for availability1 * 1 ST58,90 €Specifications:
More
Specifications
Cat. No.MANA931225ModelMANA931225PlatformColorimetric test kitsStorage temperature15...25 °CShelf life (months)24Weight98,6 g (Including packaging)