VISOCOLOR® Colorimetric Test, Cyanide
Reagents for the determination of cyanide in water samples.
- Combines simple implementation with high measurement reliability
Simple determination directly on site or in the laboratory.
- Model:MANA920128
- Platform:Colorimetric test kits
- Storage temperature:15…25 °C
- Shelf life (months):12
- Weight:205,6 g (including packaging)
Frequently Bought Together