VISOCOLOR® Colorimetric Test, Alkalinity
Supplier: MACHEREY-NAGEL
Certificates
About this item
Reagents for the determination of alkalinity in water samples.
- Combines simple implementation with high measurement reliability
Simple determination directly on site or in the laboratory.
Sea water analysis: Yes
Hazardous material: Yes
Specifications
- Model:MANA915207
- Platform:Titrimetric test kits
- Storage temperature:15...25 °C
- Shelf life (months):24
- Weight:189,4 g (Including packaging)