Spoon spatulas, micro, powder, stainless steel
: RSG
These are made of stainless steel 18/10.
- Very good chemical resistance
- Very good thermal resistance
- Quick and easy to clean
With a micro spoon at one end and a micro spatula at the other end.
Frequently Bought Together