Order Entry
Austria
ContactUsLinkComponent
Basic fuchsin for microscopical staining
Basic fuchsin for microscopical staining
Catalog # 3525.0025
Supplier:  VWR Chemicals
CAS Number:  
undefined
Basic fuchsin for microscopical staining
Catalog # 3525.0025
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Suitable for microscopy:
    Passes test

Specification Test Results

About this item

 :
 :337.85 g/mol
Melting Pt:268 °C
EINECS:209-321-2
 :GHS08
Storage Temperature:Ambient
CAS Number:632-99-5
 :58969-01-0
Formula:C₂₀H₂₀ClN₃
MDL Number:MFCD00012569