Order Entry
Austria
ContactUsLinkComponent
Calbiochem® recombinant tetanus toxoid
Calbiochem® recombinant tetanus toxoid
Certificates
Calbiochem® recombinant tetanus toxoid

Specifications

  • Protein/peptide type:
    Recombinant
  • Storage conditions:
    2...8 °C
  • Form:
    Lyophilised
  • Reconstitution instructions:
    Aliquot and freeze (–20 °C). Stock solutions are stable for up to 6 months at –20 °C
  • Protein/peptide name:
    Tetanus toxoid
  • Formulation:
    Lyophilised from 100 mm histidine, 100 mm NaCl, 5% trehalose, pH 7.0

Specifications

Frequently Bought Together