These qualitative grades of filter paper are recommended for use in analytical methods which determine or identify particulate constituents of a mixture irrespective of the amount present. Qualitative filter papers are often used in routine separation work that still requires high purity and consistent performance. These filter papers cover a wide range of laboratory applications, as liquids clarification, qualitative analytical separations for precipitates and buffers filtration, and are also used for soil analysis and in food and beverage testing. Wet-strengthened grades contain a reinforcement agent that increases their resistance to rupture when wet, such as in vacuum filtration, and their resistance when filtering acidic solutions.
Grade 5 and V5: Very fast filtering, open fiber matrix. For coarse and voluminous precipitates such as iron- aluminum- and chromium hydroxide; Si- determination in steel and pig iron analysis.
Grade 3: Fast filtering, open fiber matrix. For coarse and voluminous precipitates such as iron- aluminum- and chromium hydroxide; Si- determination in steel and pig iron analysis.
Grade V150: Medium fast filtering, open to medium fiber matrix. For qualitative tasks, coarser precipitates such as lead-, iron- and silver sulphide; alkali carbonates; calcium oxalate.
Grade 1F and V1F: Medium to slowly filtering, medium tight fiber matrix. For fast filtration of fine precipitates such as calcium oxalate, magnesium ammonium phosphate, coarser forms of barium sulphate.
Grade Munktell no. 1 and 292: Medium to slowly filtering, medium tight fiber matrix. For fast filtration of fine precipitates such as calcium oxalate, magnesium ammonium phosphate, coarser forms of barium sulphate.
Grade Munktell no. 2 and 292a: Slowly filtering, tight fiber matrix. For filtration of fine precipitates such as lead dioxide, calcium fluoride, nickel sulfide, zinc sulphide.
Grade 106: Slowly filtering for fine precipitates such as lead dioxide, calcium fluoride, nickel sulphide and zinc sulphide.
Grade 120H and V120H: Very slowly filtering, tight fiber matrix. For fine precipitates such as barium sulphate, metastannic acid and cuprous oxide.
- Catalog No:
- BINZ113304
- BINZ113314
- BINZ113427
- BINZ113428
- BINZ110128
- BINZ113368
- BINZ110100
- BINZ113415
- BINZ113411
- BINZ113410
- BINZ113409
- BINZ113407
- BINZ113404
- BINZ113377
- BINZ113344
- BINZ113376
- BINZ113306
- BINZ113370
- BINZ113389
- BINZ113388
- BINZ110112
- BINZ110109
- BINZ110101
- BINZ113384
- BINZ113382
- BINZ113336
- BINZ113338
- BINZ113341
- BINZ113366
- BINZ110098
- BINZ113364
- BINZ113334
- BINZ113362
- BINZ113414
- BINZ113327
- BINZ113323
- BINZ113317
- BINZ113423
- BINZ113421
- BINZ110127
- BINZ113390
- BINZ113346
- BINZ110113
- BINZ113348
- BINZ113425
- BINZ113417
- BINZ110125
- BINZ110010
- BINZ113391
- BINZ110122
- BINZ110124
- BINZ110123
- Grade:
- 292
- 292
- 293
- 293
- 106
- 292a
- V1F
- 293
- 293
- 293
- 293
- 293
- 293
- 292a
- 292
- 292a
- 292
- 292a
- 292a
- 292a
- V3
- V3
- V1F
- 292a
- 292a
- 292
- 292
- 292
- 292a
- V106
- 292a
- 292
- 292a
- 293
- 292
- 292
- 292
- 293
- 293
- 106
- 292a
- 292
- V3
- 292
- 293
- 293
- 106
- 106
- 292a
- 106
- 106
- 106
- Weight:
- 87 g/m²
- 87 g/m²
- 80 g/m²
- 80 g/m²
- 97 g/m²
- 80 g/m²
- 80 g/m²
- 80 g/m²
- 80 g/m²
- 80 g/m²
- 80 g/m²
- 80 g/m²
- 97 g/m²
- 87 g/m²
- 97 g/m²
- 87 g/m²
- 97 g/m²
- 97 g/m²
- 97 g/m²
- 90 g/m²
- 90 g/m²
- 80 g/m²
- 97 g/m²
- 97 g/m²
- 87 g/m²
- 87 g/m²
- 87 g/m²
- 97 g/m²
- 100 g/m²
- 97 g/m²
- 87 g/m²
- 97 g/m²
- 80 g/m²
- 87 g/m²
- 87 g/m²
- 87 g/m²
- 80 g/m²
- 80 g/m²
- 97 g/m²
- 87 g/m²
- 90 g/m²
- 87 g/m²
- 80 g/m²
- 80 g/m²
- 100 g/m²
- 97 g/m²
- Particle retention:
- 5 - 8 µm
- 5 - 8 µm
- 1 - 2 µm
- 1 - 2 µm
- 5 - 8 µm
- 5 - 6 µm
- 1 - 2 µm
- 1 - 2 µm
- 1 - 2 µm
- 1 - 2 µm
- 1 - 2 µm
- 1 - 2 µm
- 5 - 8 µm
- 5 - 8 µm
- 5 - 8 µm
- 5 - 8 µm
- 5 - 8 µm
- 5 - 8 µm
- 5 - 8 µm
- >10
- >10
- 5 - 6 µm
- 5 - 8 µm
- 5 - 8 µm
- 5 - 8 µm
- 5 - 8 µm
- 5 - 8 µm
- 5 - 8 µm
- 3 - 4 µm
- 5 - 8 µm
- 5 - 8 µm
- 5 - 8 µm
- 1 - 2 µm
- 5 - 8 µm
- 5 - 8 µm
- 5 - 8 µm
- 1 - 2 µm
- 1 - 2 µm
- 5 - 8 µm
- 5 - 8 µm
- >10
- 5 - 8 µm
- 1 - 2 µm
- 1 - 2 µm
- 3 - 4 µm
- 5 - 8 µm
- Filtration speed:
- 250 s (Herzberg)
- 250 s (Herzberg)
- 2500 s (Herzberg)
- 2500 s (Herzberg)
- 400 s (Herzberg)
- 300 s (Herzberg)
- 2500 s (Herzberg)
- 2500 s (Herzberg)
- 2500 s (Herzberg)
- 2500 s (Herzberg)
- 2500 s (Herzberg)
- 2500 s (Herzberg)
- 400 s (Herzberg)
- 250 s (Herzberg)
- 400 s (Herzberg)
- 250 s (Herzberg)
- 400 s (Herzberg)
- 400 s (Herzberg)
- 400 s (Herzberg)
- 85 s (Herzberg)
- 85 s (Herzberg)
- 300 s (Herzberg)
- 400 s (Herzberg)
- 400 s (Herzberg)
- 250 s (Herzberg)
- 250 s (Herzberg)
- 250 s (Herzberg)
- 400 s (Herzberg)
- 750 s (Herzberg)
- 400 s (Herzberg)
- 250 s (Herzberg)
- 400 s (Herzberg)
- 2500 s (Herzberg)
- 250 s (Herzberg)
- 250 s (Herzberg)
- 250 s (Herzberg)
- 2500 s (Herzberg)
- 2500 s (Herzberg)
- 400 s (Herzberg)
- 250 s (Herzberg)
- 85 s (Herzberg)
- 250 s (Herzberg)
- 2500 s (Herzberg)
- 2500 s (Herzberg)
- 750 s (Herzberg)
- 400 s (Herzberg)
- Ash content:
- 0,06%
- 0,06%
- 0,06%
- 0,06%
- 0,06%
- 0,06%
- 0,06%
- 0,06%
- 0,06%
- 0,06%
- 0,06%
- 0,06%
- 0,06%
- 0,06%
- 0,06%
- 0,06%
- 0,06%
- 0,06%
- 0,06%
- 0,06%
- 0,06%
- 0,06%
- 0,06%
- 0,06%
- 0,06%
- 0,06%
- 0,06%
- 0,06%
- 0,06%
- 0,06%
- 0,06%
- 0,06%
- 0,06%
- 0,06%
- 0,06%
- 0,06%
- 0,06%
- 0,06%
- 0,06%
- 0,06%
- 0,06%
- 0,06%
- 0,06%
- 0,06%
- 0,06%
- 0,06%