Reagent reservoir chillers
: HEATHROW
These reagent reservoir chillers are the ice-free, non-toxic way to extend the working period and preserve integrity for temperature sensitive reagents. They are made from sonic welded, engineered PP that ensures a leak free design.
- Reagents are kept at less than 5 °C for approximately 3 hours
- Ice-free solution eliminates the risk of contamination from messy ice baths
- Non-toxic blend of water and bacteriostatic agent chills and freezes quickly
- Stackable, save space when storing in the freezer or on the bench top
- Reusable, two-year warranty
The symmetrical cooler design allows reagent reservoirs to be mounted in either direction. Engineered custom fit for Heathrow Scientific 50 or 100 ml reservoirs to ensure a maximum contact for longer uniform cooling.
: Assorted packs available for color coding options.
: Not intended for autoclave, do not use with liquid nitrogen.
Frequently Bought Together