LOX assay kit (fluorometric)
Catalog # ABNOKA4191
Supplier: Abnova
CAS Number:
Specifications
- Pk:1 KIT
- Description:LOX assay kit (fluorometric)
Specifications
About this item
LOX assay kit (fluorometric) is designed for the measurement of LOX.
- Suitable sample: biological samples
- Sample volume: 50 µl
- Calibration range: 0,04 to 4 µg/ml
Delivery: The kit includes component A: amplite HRP substrate (light sensitive) (1 vial), component B: assay buffer (50 ml), component C: horseradish peroxidase (50 units) (1 vial), component D: DMSO (200 μl).
Caution: For research use only (RUO). Store the kit at –20 °C and avoid from light.