VWR®, Microcentrifuge Tubes with Flat Screw Caps
These tubes are made from ultra-clear USP Class VI PP that allows easy viewing of contents.
- RNase-, DNase- and endotoxin-free
- Engraved graduations ensure accuracy
- Fit most common rotors
- Smooth surface for applying labels
- Sterile tubes are sterilised by irradiation
These tubes feature universal screw threads for use with most popular brand screw caps. Low profile caps include a silicone rubber O-ring, ensuring a leakproof seal. (Please NOTE: Silicone O-rings have been used from June 22, 2022, first lot number: 220622626D. Manufacturer stopped using the MCT cap with ethylene propylene O-ring after June 22, 2022.)
Tubes and caps are autoclavable at 121 °C and freezable to -80 °C.
: Manufactured in a Class 100 000 cleanroom environment.
Manufactured under ISO 13485:2016 and ISO 9001:2015 quality management systems.
Certificate available for every lot.
Manufactured under ISO 13485:2016 and ISO 9001:2015 quality management systems.
Certificate available for every lot.
: Tubes are packed 500 per bag. Assorted pack of caps (525-1187) contains equal quantities of red, orange, yellow, blue and green.
- Catalog No:
- 525-1158
- 525-1157
- 525-1143
- 525-1156
- 525-1141
- 525-1144
- 525-1160
- 525-1159
- 525-1146
- 525-1145
- 525-1148
- 525-1138
- 525-1147
- 525-1137
- 525-1155
- 525-1140
- 525-1149
- 525-1139
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
Avantor Cell Culture Solutions.pdf Avantor VWR®PCR Solutions VWR Centrifuge and microcentrifuge tubes VWR Products for Nucleic Acid Preparation
Flat screw caps for microcentrifuge tubes
Frequently Bought Together