Order Entry
Austria
ContactUsLinkComponent
Ammonia solution 28.0 - 30.0% (NH₃ basis), AR Select® ACS for trace metal analysis, Macron Fine Chemicals™
Corrosive
Irritant
Environment
Ammonia solution 28.0 - 30.0% (NH₃ basis), AR Select® ACS for trace metal analysis, Macron Fine Chemicals™
Catalog # 6665-46
CAS Number:  
undefined
Corrosive
Irritant
Environment
Ammonia solution 28.0 - 30.0% (NH₃ basis), AR Select® ACS for trace metal analysis, Macron Fine Chemicals™
Catalog # 6665-46
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, clear, PVC-coated
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • ACS - Assay (as NH₃):
    28.0 - 30.0 %
  • Appearance:
    Passes Test
  • ACS - Carbon Dioxide (CO₂):
    ≤0.002 %
  • ACS - Pyridine:
    Passes Test
  • ACS - Residue after Ignition:
    ≤0.002 %
  • ACS - Substances Reducing Permanganate:
    Passes Test
  • ACS - Specific Gravity at 60°/60°F:
    0.896 - 0.902
  • Trace Impurities - Arsenic (As):
    ≤3.000 ppm
  • Chloride (Cl):
    ≤0.5 ppm
  • Trace Impurities - Heavy Metals:
    ≤0.5 ppm
  • Nitrate (NO₃):
    ≤2 ppm
  • Phosphate (PO₄):
    ≤2 ppm
  • Total Sulfur (as SO₄):
    ≤2 ppm
  • Determine by Flame Photometry & ICP - Aluminum (Al):
    ≤0.100 ppm
  • Determine by Flame Photometry & ICP - Barium (Ba):
    ≤0.100 ppm
  • Determine by Flame Photometry & ICP - Beryllium (Be):
    ≤0.010 ppm
  • Determine by Flame Photometry & ICP - Bismuth (Bi):
    ≤0.010 ppm
  • Determine by Flame Photometry & ICP - Boron (B):
    ≤0.050 ppm
  • Determine by Flame Photometry & ICP - Cadmium (Cd):
    ≤0.100 ppm
  • Determine by Flame Photometry & ICP - Calcium (Ca):
    ≤0.100 ppm
  • Determine by Flame Photometry & ICP - Chromium (Cr):
    ≤0.050 ppm
  • Determine by Flame Photometry & ICP - Cobalt (Co):
    ≤0.050 ppm
  • Determine by Flame Photometry & ICP - Copper (Cu):
    ≤0.050 ppm
  • Determine by Flame Photometry & ICP - Gallium (Ga):
    ≤0.500 ppm
  • Determine by Flame Photometry & ICP - Germanium (Ge):
    ≤0.500 ppm
  • Determine by Flame Photometry & ICP - Gold (Au):
    ≤0.500 ppm
  • Determine by Flame Photometry & ICP - Iron (Fe):
    ≤0.100 ppm
  • Determine by Flame Photometry & ICP - Lead (Pb):
    ≤0.2 ppm
  • Determine by Flame Photometry & ICP - Lithium (Li):
    ≤0.050 ppm
  • Determine by Flame Photometry & ICP - Magnesium (Mg):
    ≤0.100 ppm
  • Determine by Flame Photometry & ICP - Manganese (Mn):
    ≤0.050 ppm
  • Determine by Flame Photometry & ICP - Mercury (Hg):
    ≤0.500 ppm
  • Determine by Flame Photometry & ICP - Molybdenum (Mo):
    ≤0.010 ppm
  • Determine by Flame Photometry & ICP - Nickel (Ni):
    ≤0.500 ppm
  • Determine by Flame Photometry & ICP - Potassium (K):
    ≤0.100 ppm
  • Determine by Flame Photometry & ICP - Silicon (Si):
    ≤0.100 ppm
  • Determine by Flame Photometry & ICP - Silver (Ag):
    ≤0.050 ppm
  • Determine by Flame Photometry & ICP - Sodium (Na):
    ≤0.500 ppm
  • Determine by Flame Photometry & ICP - Strontium (Sr):
    ≤0.100 ppm
  • Determine by Flame Photometry & ICP - Thallium (Tl):
    ≤0.050 ppm
  • Determine by Flame Photometry & ICP - Tin (Sn):
    ≤0.100 ppm
  • Determine by Flame Photometry & ICP - Vanadium (V):
    ≤0.010 ppm
  • Determine by Flame Photometry & ICP - Zinc (Zn):
    ≤0.100 ppm
  • Determine by Flame Photometry & ICP - Zirconium (Zr):
    ≤0.010 ppm

Specification Test Results

About this item

 :
 :35.05 g/mol
Boiling Pt:32…37.7 °C (1013 hPa)
Melting Pt:–60…–57.5 °C
Density:0.88…0.91 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-647-6
UN:2672
ADR:8,III
 :Management of Change (MOC) category = N
 :GHS05
 :GHS07
 :GHS09
CAS Number:1336-21-6
 :7664-41-7
Formula:NH₃
MDL Number:MFCD00066650