Order Entry
Austria
ContactUsLinkComponent
NWTPH-HCID surrogate standard, certified reference material, Supelco®
NWTPH-HCID surrogate standard, certified reference material, Supelco®
Catalog # UST113-1.5ML
Supplier:  Merck
CAS Number:  
NWTPH-HCID surrogate standard, certified reference material, Supelco®
Catalog # UST113-1.5ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    5000 µg/ml in methylene chloride
  • Pk:
    1,5 mL
  • Description:
    NWTPH-HCID surrogate standard

Specifications

About this item