Order Entry
Austria
ContactUsLinkComponent
Supelco® Organic Reference Standard, ASTM® D6584 Standard Solution Kit
Supelco® Organic Reference Standard, ASTM® D6584 Standard Solution Kit
Catalog # CRM44918
Supplier:  Merck
CAS Number:  
Supelco® Organic Reference Standard, ASTM® D6584 Standard Solution Kit
Catalog # CRM44918
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1
  • Standards:
    With Internal Standards, certified reference material
  • Description:
    ASTM® D6584 Standard Solution Kit
  • Storage conditions:
    2…8 °C
  • Hazard class description:
    H225 - H302 + H312 + H332 - H315 - H319
  • Shipping conditions:
    WICE
  • M-Clarity quality level:
    MQ100
  • Flash point:
    17,0 °C - Closed cup

Specifications

About this item

  • GHS signal: GHS02, GHS07
Caution: P210 - P280 - P301 + P312 - P303 + P361 + P353 - P304 + P340 + P312 - P305 + P351 + P338