Supelco® Organic Reference Standard, Aromatic Hydrocarbon Standard
UST122-1.5ML
: Merck
:
- Concentration:1000 µg/ml in methylene chloride
- Pk:1,5 mL
- Standards:Certified reference material
- Description:Aromatic hydrocarbon standard
- Storage conditions:2...30 °C
- Hazard class description:H315 - H319 - H336 - H350 - H410
- Shipping conditions:STD
- M-Clarity quality level:MQ100
- GHS signal: GHS08, GHS07, GHS09
: P202 - P261 - P273 - P302 + P352 - P305 + P351 + P338 - P308 + P313