Order Entry
Austria
ContactUsLinkComponent
 
Zinc oxide ≥99.0% (by complexometric analysis), Puriss. p.a. ACS, Fluka™
  96479-500G
 :  
Zinc oxide ≥99.0% (by complexometric analysis), Puriss. p.a. ACS, Fluka™
  96479-500G
 :  

 

  • Pack type:
    Plastic bottle
  • Pk:
    500 g
  • Seq:
    3645

 

 

  • Assay:
    99.0 - 101.0 %
  • Insoluble in acid:
    Max. 0.01 %
  • Alkalinely reacting substances:
    complying
  • Arsenic (As):
    Max. 1 ppm
  • Calcium (Ca):
    Max. 10 ppm
  • Cadmium (Cd):
    Max. 5 ppm
  • Cobalt (Co):
    Max. 5 ppm
  • Chromium (Cr):
    Max. 5 ppm
  • Copper (Cu):
    Max. 5 ppm
  • Iron (Fe):
    Max. 5 ppm
  • Potassium (K):
    Max. 50 ppm
  • Magnesium (Mg):
    Max. 5 ppm
  • Manganese (Mn):
    Max. 5 ppm
  • Sodium (Na):
    Max. 10 ppm
  • Nickel (Ni):
    Max. 5 ppm
  • Lead (Pb):
    Max. 20 ppm
  • Chloride (Cl):
    Max. 10 ppm
  • Nitrate (NO₃):
    Max. 30 ppm
  • Total S (as SO₄):
    Max. 100 ppm

 

 

 :
 :81.39 g/mol
 :13,10200
 :1.975 °C
 :5.6 g/cm³ (20 °C)
 :Ambient
 :215-222-5
 :01-2119463881-32
 :1314-13-2
 :ZnO
 :MFCD00011300