Order Entry
Austria
ContactUsLinkComponent
Water with 0,1% (v/v) acetic acid, CHROMASOLV™ LC-MS Ultra LabReady™, Solvent Blend tested for UHPLC-MS, Riedel-de Haën™
Water with 0,1% (v/v) acetic acid, CHROMASOLV™ LC-MS Ultra LabReady™, Solvent Blend tested for UHPLC-MS, Riedel-de Haën™
Catalog # 14282-2L
CAS Number:  
Water with 0,1% (v/v) acetic acid, CHROMASOLV™ LC-MS Ultra LabReady™, Solvent Blend tested for UHPLC-MS, Riedel-de Haën™
Catalog # 14282-2L
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2 L
  • Seq:
    5086

Specifications

Specification Test Results

  • Assay of acetic acid:
    0.093 - 0.107 %
  • pH (20 °C):
    3.2 - 3.5
  • Calcium (Ca):
    Max. 0.2 ppm
  • Potassium (K):
    Max. 0.2 ppm
  • Magnesium (Mg):
    Max. 0.2 ppm
  • Sodium (Na):
    Max. 0.5 ppm
  • Transmittance at 254 nm:
    Min. 99 %
  • Transmittance at 230 nm:
    Min. 75 %
  • Transmittance at 210 nm:
    Min. 20 %
  • UHPLC-MS ESI(+) (Reserpine):
    Max. 10 ppb
  • UHPLC-MS ESI(-) (Digoxin):
    Max. 50 ppb
  • Suitability for the UHPLC-MS:
    complying
  • LC-MS application:
    complying
  • HPLC Gradient at 254 nm:
    Max. 2 mAU
  • HPLC Gradient at 210 nm:
    Max. 20 mAU

Specification Test Results

About this item

 :
Density:1.000 g/cm³
Storage Temperature:Ambient
EINECS:231-791-2
 :7732-18-5
MDL Number:MFCD00011332