


Tetramethylammonium hydroxide 0.1 M (CH3)4NOH (0.1N) in 2-propanol/methanol volumetric solution, for titration in non-aqueous media, Fluka™
Catalog # 35436-1L
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Factor:0.999 - 1.001
Specification Test Results
About this item
:
:91.15 g/mol
Flash Pt:12 °C
Storage Temperature:Ambient
UN:1992
ADR:3,II
:GHS08
:GHS06
:GHS02
CAS Number:75-59-2
Formula:C₄H₁₃NO
MDL Number:MFCD00008280