

Tashiro’s indicator solution in methanol, Fluka™
Catalog # 35325-1L
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Change range:complying
Specification Test Results
About this item
:
Flash Pt:18 °C
Storage Temperature:Ambient
UN:1230
ADR:3,II
:GHS02
:GHS08
MDL Number:MFCD00148563