Silica gel Rubin drying bags, beads 1.0-3.0 mm, with indicator 1/6 TE ( ~5 g/bag), Fluka™
72811-1KG
:
- Pack type:Plastic bottle
- Pk:1 kg
- Seq:4024
- Form:beads
- Appearance:complying
- Water absorbing capacity:Min. 20 %
- Change:complying
- Sieve analysis 1-3 mm:Min. 95 %
:
:60.08 g/mol
:2.230 °C
:1.600 °C
:2 g/cm³ (20 °C)
:Ambient
:231-545-4
:112926-00-8
:63231-67-4
:SiO₂
:MFCD00011232