Order Entry
Austria
ContactUsLinkComponent
Magnesium sulphate concentrate for dilution to 1L, 0.1 M MgSO4 volumetric solution, Fluka™
Magnesium sulphate concentrate for dilution to 1L, 0.1 M MgSO4 volumetric solution, Fluka™
Catalog # 38146-1EA
CAS Number:  
undefined
Magnesium sulphate concentrate for dilution to 1L, 0.1 M MgSO4 volumetric solution, Fluka™
Catalog # 38146-1EA
CAS Number:  

Specifications

  • Pack type:
    Ampoule
  • Pk:
    1
  • Seq:
    0001

Specifications

Specification Test Results

  • Factor:
    0.998 - 1.002

Specification Test Results

About this item

 :
 :120.37 g/mol
Storage Temperature:Ambient
CAS Number:7487-88-9
Formula:MgSO₄
MDL Number:MFCD00011110