Iodine 0.025 M I2 (0.05N) volumetric solution, Fluka™
Catalog # 405353-500ML
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 mL
- Seq:2725
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:0.0480 - 0.0520 N
Specification Test Results
About this item
:
:253.81 g/mol
Density:1.03 g/cm³ (25 °C)
:GHS08
Storage Temperature:Ambient
CAS Number:7553-56-2
Formula:I₂
MDL Number:MFCD00011355