Order Entry
Austria
ContactUsLinkComponent
Hydrochloric acid 34 - 37%, TraceSELECT™ for inorganic trace analysis, Fluka™
Corrosive
Irritant
Hydrochloric acid 34 - 37%, TraceSELECT™ for inorganic trace analysis, Fluka™
Catalog # 72787-2.5L
CAS Number:  
Corrosive
Irritant
Hydrochloric acid 34 - 37%, TraceSELECT™ for inorganic trace analysis, Fluka™
Catalog # 72787-2.5L
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Assay:
    34 - 37 %
  • Silver (Ag):
    Max. 0.5 ppb
  • Aluminium (Al):
    Max. 5 ppb
  • Arsenic (As):
    Max. 5 ppb
  • Gold (Au):
    Max. 1 ppb
  • Barium (Ba):
    Max. 0.5 ppb
  • Beryllium (Be):
    Max. 0.5 ppb
  • Bismuth (Bi):
    Max. 0.5 ppb
  • Calcium (Ca):
    Max. 10 ppb
  • Cadmium (Cd):
    Max. 0.5 ppb
  • Cobalt (Co):
    Max. 0.5 ppb
  • Chromium (Cr):
    Max. 1 ppb
  • Cesium (Cs):
    Max. 0.5 ppb
  • Copper (Cu):
    Max. 1 ppb
  • Iron (Fe):
    Max. 5 ppb
  • Gallium (Ga):
    Max. 0.5 ppb
  • Mercury (Hg):
    Max. 0.5 ppb
  • Indium (In):
    Max. 0.5 ppb
  • Potassium (K):
    Max. 10 ppb
  • Lithium (Li):
    Max. 0.5 ppb
  • Magnesium (Mg):
    Max. 1 ppb
  • Manganese (Mn):
    Max. 0.5 ppb
  • Molybdenum (Mo):
    Max. 0.5 ppb
  • Sodium (Na):
    Max. 10 ppb
  • Nickel (Ni):
    Max. 1 ppb
  • Lead (Pb):
    Max. 0.5 ppb
  • Palladium (Pd):
    Max. 1 ppb
  • Platinum (Pt):
    Max. 0.5 ppb
  • Rubidium (Rb):
    Max. 0.5 ppb
  • Rhodium (Rh):
    Max. 0.5 ppb
  • Ruthenium (Ru):
    Max. 0.5 ppb
  • Antimony (Sb):
    Max. 0.5 ppb
  • Selenium (Se):
    Max. 1 ppb
  • Tin (Sn):
    Max. 1 ppb
  • Strontium (Sr):
    Max. 0.5 ppb
  • Titanium (Ti):
    Max. 1 ppb
  • Thallium (Tl):
    Max. 0.5 ppb
  • Uran (U):
    Max. 0.5 ppb
  • Vanadium (V):
    Max. 1 ppb
  • Zinc (Zn):
    Max. 1 ppb
  • Zirconium (Zr):
    Max. 0.5 ppb
  • Free chlorine (Cl):
    Max. 0.5 ppm
  • Bromide (Br):
    Max. 10000 ppb
  • Phosphate (PO₄):
    Max. 50 ppb
  • Sulfate (SO₄):
    Max. 1000 ppb
  • Vapor Pressure:
    7.93 psi ( 37.7 °C)

Specification Test Results

About this item

 :
Boiling Pt:42 °C (1.013 hPa)
Melting Pt:–35 °C
Density:1.190 g/cm³ (20 °C)
Storage Temperature:Ambient
ADR:8,II
 :36.46 g/mol
UN:1789
 :GHS05
 :GHS07
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324