Order Entry
Austria
ContactUsLinkComponent
Ammonia solution ≥25% (NH₃ basis) in water, TraceSELECT™ for trace analysis, Fluka™
Irritant
Corrosive
Ammonia solution ≥25% (NH₃ basis) in water, TraceSELECT™ for trace analysis, Fluka™
Catalog # 09857-500ML
CAS Number:  
Irritant
Corrosive
Ammonia solution ≥25% (NH₃ basis) in water, TraceSELECT™ for trace analysis, Fluka™
Catalog # 09857-500ML
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

Specification Test Results

  • Assay of NH₃:
    25.0 - 31.0 %
  • Density (D 20/4):
    0.890 - 0.910
  • Silver (Ag):
    Max. 0.5 ppb
  • Aluminium (Al):
    Max. 5 ppb
  • Arsenic (As):
    Max. 1 ppb
  • Gold (Au):
    Max. 0.5 ppb
  • Barium (Ba):
    Max. 0.5 ppb
  • Beryllium (Be):
    Max. 0.5 ppb
  • Bismuth (Bi):
    Max. 0.5 ppb
  • Calcium (Ca):
    Max. 10 ppb
  • Cadmium (Cd):
    Max. 0.5 ppb
  • Cerium (Ce):
    Max. 0.5 ppb
  • Cobalt (Co):
    Max. 0.5 ppb
  • Chromium (Cr):
    Max. 1 ppb
  • Cesium (Cs):
    Max. 0.5 ppb
  • Copper (Cu):
    Max. 0.5 ppb
  • Iron (Fe):
    Max. 2 ppb
  • Gallium (Ga):
    Max. 0.5 ppb
  • Germanium (Ge):
    Max. 0.5 ppb
  • Mercury (Hg):
    Max. 1 ppb
  • Indium (In):
    Max. 0.5 ppb
  • Iridium (Ir):
    Max. 0.5 ppb
  • Potassium (K):
    Max. 5 ppb
  • Lithium (Li):
    Max. 0.5 ppb
  • Magnesium (Mg):
    Max. 1 ppb
  • Manganese (Mn):
    Max. 0.5 ppb
  • Molybdenum (Mo):
    Max. 0.5 ppb
  • Sodium (Na):
    Max. 10 ppb
  • Nickel (Ni):
    Max. 1 ppb
  • Lead (Pb):
    Max. 0.5 ppb
  • Palladium (Pd):
    Max. 0.5 ppb
  • Platinum (Pt):
    Max. 0.5 ppb
  • Rubidium (Rb):
    Max. 0.5 ppb
  • Rhodium (Rh):
    Max. 0.5 ppb
  • Ruthenium (Ru):
    Max. 0.5 ppb
  • Antimony (Sb):
    Max. 0.5 ppb
  • Selenium (Se):
    Max. 1 ppb
  • Tin (Sn):
    Max. 0.5 ppb
  • Strontium (Sr):
    Max. 0.5 ppb
  • Titanium (Ti):
    Max. 1 ppb
  • Thallium (Tl):
    Max. 0.5 ppb
  • Uran (U):
    Max. 0.5 ppb
  • Vanadium (V):
    Max. 1 ppb
  • Zinc (Zn):
    Max. 1 ppb
  • Zirconium (Zr):
    Max. 0.5 ppb
  • Carbonate (as CO₂):
    Max. 10 ppm
  • Bromide (Br):
    Max. 50 ppb
  • Chloride (Cl):
    Max. 500 ppb
  • Fluoride (F):
    Max. 100 ppb
  • Iodide (I):
    Max. 50 ppb
  • Nitrite (NO₂):
    Max. 100 ppb
  • Nitrate (NO₃):
    Max. 500 ppb
  • Phosphate (PO₄):
    Max. 50 ppb
  • Sulfate (SO₄):
    Max. 100 ppb

Specification Test Results

About this item

 :
 :35.05 g/mol
Boiling Pt:37.7 °C (1.013 hPa)
Melting Pt:-92 °C
Density:0.90 g/cm³ (20 °C)
UN:2672
ADR:8,III
 :GHS07
 :GHS05
Storage Temperature:Ambient
CAS Number:1336-21-6
 :7664-41-7
Formula:NH₃
MDL Number:MFCD00066650
Melting Pt:–92 °C