Order Entry
Austria
ContactUsLinkComponent
 
 
 
Potassium pyroantimonate ≥99.0%, AnalaR NORMAPUR® analytical reagent
  86173.180
 :  
undefined
 
 
Potassium pyroantimonate ≥99.0%, AnalaR NORMAPUR® analytical reagent
  86173.180
 :  

 

  • Pack type:
    Plastic bottle
  • Pk:
    100 g
  • Seq:
    3401

 

 

  • Appearance:
    white powder
  • Assay (as KSbO₃):
    min. 99,0%
  • Loss on ignition (600 °C):
    max. 22%
  • Free alkali (as KOH):
    max. 0,8%
  • Sodium (Na):
    max. 0,2%

 

 

 :
 :262.9 g/mol
 :1549
 :6.1,III
 :GHS07
 :GHS09
 :12208-13-8
 :MFCD00150439
 :KSbO₃·3H₂O
 :MFCD00011415