



Glutaraldehyd 25% in wässriger Lösung, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:111-30-8
:
Formula:C₅H₈O₂
MDL Number:MFCD00007025
:100.12 g/mol
Melting Pt:–6 °C
Storage Temperature:Kühlschrank
UN:2922
ADR:8 (6.1),II
:GHS06
:GHS05
:GHS08
:GHS09
Specification Test Results
- Appearance:clear colourless or yellow liquid
- Assay:23 - 27 %
- Color scale:max. 50 APHA
- Density (d20 °C/4 °C):1,055 - 1,065
- pH:3,2 - 4,2
- Methanol:max.0,03%
Specification Test Results
Related Information
Certificates
Search by Lot Numberspec_VWRC20873.180.pdf spec_VWRC20873.290.pdf spec_VWRC20873.320.pdf spec_VWRC20873.460
Product specification
Frequently Bought Together