Order Entry
Austria
ContactUsLinkComponent
 
 
Magnesium 2% in nitric acid 5% ≥99.9995%, TraceSELECT™ matrix modifier solution, Fluka™
  428892-100ML
 :  
 
Magnesium 2% in nitric acid 5% ≥99.9995%, TraceSELECT™ matrix modifier solution, Fluka™
  428892-100ML
 :  

 

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Seq:
    2440

 

 

  • Assay of Mg:
    1.5 - 2.5 %
  • Nitric acid:
    1.0 - 5.0 %
  • Metal trace analysis (ICP):
    Max. 5.5 ppm

 

 

 :
 :24.3 g/mol
 :1.093 g/cm³ (25 °C)
 :Ambient
 :3264
 :8,III
 :GHS05
 :7439-95-4
 :Mg
 :MFCD00085308