ContactUsLinkComponent
Test strips, Total sugar, QUANTOFIX®
Test strips, Total sugar, QUANTOFIX®
Catalog # MANA91352
Supplier:  MACHEREY-NAGEL
CAS Number:  
undefined
Test strips, Total sugar, QUANTOFIX®
Catalog # MANA91352
Supplier:  MACHEREY-NAGEL
CAS Number:  
Specifications
  • Chemical method:
    Enzymatic reaction
  • Pk:
    100 Tests
  • Description:
    Total sugar test strips, yellow - ochre
  • Range:
    0 - 800 mg/l fructose/glucose
Specifications
About this item
QUANTOFIX® test strips meet all requirements of a modern rapid test. The colour of the reactive pad changes depending on the concentration of an analyte in the sample. The evaluation is usually carried out visually by comparison of the reaction colour with a multi-stage colour scale. The test strips are intended for single use, maintenance or calibration are not required.
  • Results within seconds: Just dip, wait and read against multi-stage colour scale
  • Always ready to use: No accessories necessary for evaluation
  • Safe results: Desiccant in the stopper for optimal protection of the strips against humidity
These test strips allow the quick and easy determination of the monosaccharides fructose and glucose in solutions. The easy procedure provides reliable results in 10 minutes. The determination of the fructose and glucose is frequently used in food industry (especially potatoes, honey, spinach and many more) and beverage industry (especially fruit juice, wine and many more).
Gradation: 0 · 55 · 100 · 250 · 400 · 600 · 800 mg/l fructose/glucose