Order Entry
Austria
ContactUsLinkComponent
Test strips, Glutaraldehyde, QUANTOFIX®
Test strips, Glutaraldehyde, QUANTOFIX®
Catalog # 705-1159
Supplier:  MACHEREY-NAGEL
CAS Number:  
undefined
Test strips, Glutaraldehyde, QUANTOFIX®
Catalog # 705-1159
Supplier:  MACHEREY-NAGEL
CAS Number:  

Specifications

  • Chemical method:
    Mixed indicator
  • Pk:
    100 Tests
  • Description:
    Glutaraldehyde test strips, bright orange - magenta
  • Range:
    0 - 2,5 % glutaraldehyde

Specifications

About this item

QUANTOFIX® test strips meet all requirements of a modern rapid test. The colour of the reactive pad changes depending on the concentration of an analyte in the sample. The evaluation is usually carried out visually by comparison of the reaction colour with a multi-stage colour scale. The test strips are intended for single use, maintenance or calibration are not required.

  • Results within seconds: Just dip, wait and read against multi-stage colour scale
  • Always ready to use: No accessories necessary for evaluation
  • Safe results: Desiccant in the stopper for optimal protection of the strips against humidity

The test strips allow the fast and reliable determination of glutaraldehyde in solutions. The easy dip-and-read procedure provides reliable results in 20 seconds. Glutaraldehyde is a strong disinfectant. It is, for example, often used in healthcare and medical engineering to disinfect surgical instruments and equipment. Overall, glutaraldehyde is one of the most popular disinfectants in single-use device reprocessing.These test strips are to test the glutaraldehyde concentration in solutions up to 2,5%.

Certifications: CE marked. These test strips conform to the Medical Device Directive 93/42/EEC.