Order Entry
Austria
Orders LinkContactUsLinkComponent
Test strips, Calcium, QUANTOFIX® | Avantor

Test strips, Calcium, QUANTOFIX®

QUANTOFIX Calcium, 0 • 10 • 25 • 50 • 100 - mg/L - Ca²⁺, Complexation with glyoxal-bis(2-hydroxyanil), 148×122×40 mm (package), 60 test(s), 100 Tests
Catalog # MANA91324
Supplier:   MACHEREY-NAGEL
Test strips, Calcium, QUANTOFIX®
Catalog # MANA91324
Supplier:  MACHEREY-NAGEL
CAS Number:  
Test strips, Calcium, QUANTOFIX®
Test strips, Calcium, QUANTOFIX®

QUANTOFIX Calcium, 0 • 10 • 25 • 50 • 100 - mg/L - Ca²⁺, Complexation with glyoxal-bis(2-hydroxyanil), 148×122×40 mm (package), 60 test(s), 100 Tests

CAS Number:  
Price
69,60 €
Log in for availability
1 * 100 Tests
Product Details & Documents

QUANTOFIX® test strips meet all requirements of a modern rapid test. The colour of the reactive pad changes depending on the concentration of an analyte in the sample. The evaluation is usually carried out visually by comparison of the reaction colour with a multi-stage colour scale. The test strips are intended for single use, maintenance or calibration are not required.

  • Results within seconds: Just dip, wait and read against multi-stage colour scale
  • Always ready to use: No accessories necessary for evaluation
  • Safe results: Desiccant in the stopper for optimal protection of the strips against humidity

The test strips allow the quick and easy determination of calcium in solutions. They come with all necessary reagents so that the measurement can immediately be started. Within one minute reliable results are obtained. Calcium is an important element in nutrition and is therefore tested in foodstuff. It is essential for the building of human bones. Lack of calcium in the daily diet can lead to osteoporosis. The recommended consumption is about 1000 mg/day. In combination with magnesium, calcium is responsible for water hardness.

Gradation: 0 · 10 · 25 · 50 · 100 mg/l Ca2+

Specifications

Specifications

  • Chemical method:
    Complexation with glyoxal-bis(2-hydroxyanil)
  • Pk:
    100 Tests
  • Description:
    QUANTOFIX Calcium
  • Dimensions:
    148×122×40 mm (package)
  • Tests per kit:
    60 test(s)
  • Range:
    0 • 10 • 25 • 50 • 100 - mg/L - Ca²⁺
  • Cat. No.:
    MANA91324
  • Parameters:
    Requires additional reagent
  • Contents:
    60 test strips in a tube, additional regaents, instruction leaflet in a box
  • Platform:
    Semi-quantitative test strips
  • Compliance:
    Hazardous material
  • Instrument:
    No
  • Shelf life (months):
    30 (from production)
  • Method:
    Calcium - Glyoxal-bis(2-hydroxyaniline) (yellow → red)
  • Weight:
    186 g

Specifications

Empfehlungen

Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.

Andernfalls erhalten Sie allgemeine Empfehlungen.